missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Cell Culture Dishes, Non-Treated
For cell based assays and microscopic analysis. Eppendorf™ Cell Culture Dishes, Non-Treated helps in direct access to the cells.
Brand: Eppendorf™ 0030701011
This item is not returnable.
View return policy
Description
- Safety in handling during transportation and while working due to corrugated handling ring
- SplashProtect ring inside of the dish lid traps liquid and prevents spills during transportation or incubation
- Pronounced rim on dish lid and exact fitting of lid and bottom ensure sturdy, secure stacking
- Easy differentiation of lid and dish bottom prevents unintended removal of the lid
- Packaging combines tool-free opening, resealable top and side for tape-free closing, and an easy solution to shrink the package for space-conscious storage
- Available in three formats: 35, 60 and 100mm
Specifications
Transparent | |
Polystyrene | |
Flat | |
Transparent | |
Polystyrene | |
Sterile | |
Cell Culture Dish | |
Lid |
Closure Included | |
Not Autoclavable | |
26.6 mL | |
21.9 sq. cm | |
Round | |
Non-treated | |
3 to 5 mL | |
14 mm |