Learn More
Invitrogen™ eBioscience™ Cell Stimulation Cocktail (500X)
Cocktail of phorbol 12-myristate 13-acetate (PMA) and ionomycin. Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines.
984.00 SEK - 2800.00 SEK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15452024
|
Invitrogen™
00-4970-93 |
100 μL |
984.00 SEK
100µL |
In Stock
Log in to see stock. |
|||||
15290035
|
Invitrogen™
00-4970-03 |
4 x 100 μL |
2800.00 SEK
400µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Can be used to induce cytokine production in in vitro cell culture for subsequent detection in immunoassays
- Can also be used in conjunction with the Protein Transport Inhibitor Cocktail (500X) (cat. 00-4980) for intracellular retention of the induced cytokine expression
Specifications
eBioscience™ | |
Cell Stimulation Cocktail |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- eBioscience Cell Stimulation Cocktail (500X)
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol