Learn More
Eppendorf™ Centrifuge Adapters for Plate Rotors
For use with plate rotors.
Brand: Eppendorf™ 5825711009
1907.60 SEK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
This item is not returnable.
View return policy
Description
- Available in either 96- or 384-well PCR plates
- Supplied in sets of 2
Specifications
Adapter | |
96-Well PCR Plate | |
4 |
PCR Plate | |
all MTP/DWP Rotor Buckets | |
2 Pack |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%
Product Content Correction
Product Title
Eppendorf™ Centrifuge Adapters for Plate Rotors > For 96-Well PCR Plate
Spot an opportunity for improvement?Share a Content Correction