missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ ChromatoPur™ Bovine Thrombin, Low Specific Activity, Diagnostic Grade

Suitable as a diagnostic and research reagent in clotting assays

Brand:  MP Biomedicals™ 0219990780


 View more versions of this product

Product Code. 15722422

  • 6205.00 SEK / 100ku

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Chromatographically purified with minimal loss of native clotting activity and minimized degradation to the gamma form. Enzyme activity is measured by clotting assay. 90 to 300U/mg protein.

Physical Characteristics

  • Appearance: White to off-white powder
  • pH: 7.0 to 8.0
  • Protein: ≥50%
  • Specific activity: 90 to 300U/mg protein
  • Activity: Report μ/mg solid
  • Total Activity: U/vial
  • Exipients: Reported
Certifications: ISO, cGMP

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cell Culture, Microbial Culture
7 to 8
Bovine Thrombin
Solid
Bovine Thrombin
100 kU
-10°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Bovine Thrombin, Low Specific Activity, Diagnostic Use

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Thrombin

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.