missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ ChromatoPur™ Bovine Thrombin, Low Specific Activity, Diagnostic Grade
Suitable as a diagnostic and research reagent in clotting assays
1750.00 SEK - 6205.00 SEK
Specifications
For Use With (Application) | Cell Culture, Microbial Culture |
---|---|
Name | Bovine Thrombin |
pH Range | 7 to 8 |
Source | Bovine Thrombin |
Storage Requirements | -10°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15722422
|
MP Biomedicals™
0219990780 |
100 kU |
6205.00 SEK
100ku |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15712422
|
MP Biomedicals™
0219990725 |
25 kU |
2715.00 SEK
25ku |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11461665
|
MP Biomedicals™
0219990710 |
10 kU |
1750.00 SEK
10ku |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chromatographically purified with minimal loss of native clotting activity and minimized degradation to the gamma form. Enzyme activity is measured by clotting assay. 90 to 300U/mg protein.
Physical Characteristics
- Appearance: White to off-white powder
- pH: 7.0 to 8.0
- Protein: ≥50%
- Specific activity: 90 to 300U/mg protein
- Activity: Report μ/mg solid
- Total Activity: U/vial
- Exipients: Reported
Specifications
Cell Culture, Microbial Culture | |
7 to 8 | |
-10°C |
Bovine Thrombin | |
Bovine Thrombin | |
Solid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Bovine Thrombin, Low Specific Activity, Diagnostic Use
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Thrombin
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title