missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek Spot-Label™ Alexa Fluor™ 488
Spot VHH conjugated to Alexa Fluor™ 488.
2025.00 SEK - 4310.00 SEK
Specifications
Clonality | Monoclonal |
---|---|
For Use With (Application) | Immunofluorescence (IF), Western Blot |
Conjugate | Alexa Fluor™ 488 |
Target Specificity | Spot-Tag (PDRVRAVSHWSS) |
Content And Storage | Shipped at ambient temperature. Upon receipt store at +4°C / 40°F.Stable for 6 months. Do not freeze. Protect from light. |
Description
- anti-Spot-Tag VHH/ Nanobody conjugated to fluorescent dye for immunofluorescence, microscopy, and immunoblotting of Spot-tagged proteins
Specifications
Monoclonal | |
Alexa Fluor™ 488 | |
Shipped at ambient temperature. Upon receipt store at +4°C / 40°F.Stable for 6 months. Do not freeze. Protect from light. |
Immunofluorescence (IF), Western Blot | |
Spot-Tag (PDRVRAVSHWSS) |