Learn More
Hirschmann™ Class A PMP Volumetric Flask with Stopper
Calibrated to contain a precise volume at a specific temperature. Flasks are used for precise dilutions and prepare standard solutions
Brand: Hirschmann™ 7570186
This item is not returnable.
View return policy
Description
- Acc. to DIN EN ISO 1042
- PMP (TPX)
- Adjusted ringmark
- Translucent
- With standard ground poly stoppers
Specifications
DIN EN ISO 1042 | |
250 mL | |
5/Pack | |
A | |
Stopper |
PMP | |
235 mm | |
± 0.15 mL | |
Round |