Learn More
Hirschmann™ Class A PMP Volumetric Flask with Stopper
Calibrated to contain a precise volume at a specific temperature. Flasks are used for precise dilutions and prepare standard solutions
Brand:  Hirschmann™ 7570170
 View more versions of this product
Product Code. 13158069
  • 1097.75 SEK / Pack of 6
Explore more special offers
Description
Description
  • Acc. to DIN EN ISO 1042
  • PMP (TPX)
  • Adjusted ringmark
  • Translucent
  • With standard ground poly stoppers
Specifications
Specifications
DIN EN ISO 1042
25 mL
6/Pack
A
Stopper
PMP
115 mm
± 0.04 mL
Round
NS 10/19