Learn More
Hirschmann™ Class A PMP Volumetric Flask with Stopper
Calibrated to contain a precise volume at a specific temperature. Flasks are used for precise dilutions and prepare standard solutions
Brand:  Hirschmann™ 7570190
 View more versions of this product
Product Code. 13198069
  • 1398.37 SEK / Pack of 4
Explore more special offers
Description
Description
  • Acc. to DIN EN ISO 1042
  • PMP (TPX)
  • Adjusted ringmark
  • Translucent
  • With standard ground poly stoppers
Specifications
Specifications
DIN EN ISO 1042
500 mL
4/Pack
A
Stopper
PMP
270 mm
± 0.25 mL
Round
NS 19/26