missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNTNAP5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
1732.00 SEK - 4375.00 SEK
Specifications
Antigen | CNTNAP5 |
---|---|
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17723484
|
Invitrogen
PA5122084 |
20 μL |
1732.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17713484
|
Invitrogen
PA5122083 |
100 μL |
4375.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May play a role in the correct development and proper functioning of the peripheral and central nervous system and be involved in cell adhesion and intercellular communication.Specifications
CNTNAP5 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q8WYK1 | |
129684 | |
Ala25-Asp302 | |
Primary | |
-20°C | |
CNTNAP5 |
1 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
CASPR5; cell recognition molecule Caspr5; CNTNAP5; contactin associated protein like 5; contactin associated protein-like 5; contactin-associated protein-like 5 | |
CNTNAP5 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title