missing translation for 'onlineSavingsMsg'
Learn More

COD Reagent, Chem-Lab

Determine the amount of pollutant in a water sample. COD Reagent, Chem-Lab is an oxidizing agent for COD analysis, comprised of mercury sulfate with potassium dichromate in sulfuric acid. The solution meets UNE 77-004-89 standards and can be used in AFNOR-validated analyses of water quality.

Brand:  Chem Lab CL02.0329.1000


 View more versions of this product

Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery. Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11919192

  • 738.00 SEK / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Chemical oxygen demand (COD) is a measure of the amount of oxygen that can be consumed by reactions in a measured solution. A COD test quantifies the amount of organic materials in waste water or surface water. COD measurements are commonly made on samples to determine the amount of oxidizable pollutants and to determine the effect a particular effluent will have on a receiving body.

  • According to UNE 77-004-89 standards
  • For AFNOR-validated determinations of water samples
  • 80g HgSO4 + 11.7685g K2Cr2O7 per 1L 10% H2SO4
    Specifications

    Specifications

    UNE 77-004-89, AFNOR
    COD Reagent
    1 L
    80g HgSO4 + 11.7685g K2Cr2O7/ 1L 10% H2SO4 = 0.24 N
    COD water determination
    Product Suggestions

    Product Suggestions

    Videos
    Safety and Handling

    Safety and Handling

    Product Identifier
    • COD reagent acc. UNE 77-004-89

    Signal Word
    • Danger

    Hazard Category
    • Acute toxicity Category 3
    • LONG-TERM AQUATIC HAZARD Chronic 1
    • Carcinogenicity Category 1
    • Substances and mixtures corrosive to metals Category 1
    • Germ cell mutagenicity Category 1
    • Reproductive toxicity Category 1
    • Skin corrosion/irritation Category 1 A
    • Specific target organ toxicity after repeated exposure Category 2

    Hazard Statement
    • H290-May be corrosive to metals.
    • H301-Toxic if swallowed.
    • H314-Causes severe skin burns and eye damage.
    • H340-May cause genetic defects.
    • H350-May cause cancer.
    • H360-May damage fertility or the unborn child.
    • H373-May cause damage to organs.
    • H410-Very toxic to aquatic life with long lasting effects.

    Precautionary Statement
    • P273-Avoid release to the environment.
    • P280-Wear protective gloves/protective clothing/eye protection/face protection.
    • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
    • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
    • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
    • P310-Immediately call a POISON CENTER/doctor/.

    Supplemental information
    • MIXTURE LIST-Contains : mercury(II) sulphate, potassium dichromate, sulphuric acid

    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.