Learn More
COD Reagent, Chem-Lab
Determine the amount of pollutant in a water sample. COD Reagent, Chem-Lab is an oxidizing agent for COD analysis, comprised of mercury sulfate with potassium dichromate in sulfuric acid. The solution meets UNE 77-004-89 standards and can be used in AFNOR-validated analyses of water quality.
738.00 SEK - 1665.00 SEK
Specifications
Certifications/Compliance | UNE 77-004-89, AFNOR |
---|---|
Composition | 80g HgSO4 + 11.7685g K2Cr2O7/ 1L 10% H2SO4 = 0.24 N |
Description | COD Reagent |
For Use With (Application) | COD water determination |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11919192
|
Chem Lab
CL02.0329.1000 |
1 L |
738.00 SEK
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11957107
|
Chem Lab
CL02.0329.2500 |
2.5 L |
1665.00 SEK
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical oxygen demand (COD) is a measure of the amount of oxygen that can be consumed by reactions in a measured solution. A COD test quantifies the amount of organic materials in waste water or surface water. COD measurements are commonly made on samples to determine the amount of oxidizable pollutants and to determine the effect a particular effluent will have on a receiving body.
Specifications
UNE 77-004-89, AFNOR | |
80g HgSO4 + 11.7685g K2Cr2O7/ 1L 10% H2SO4 = 0.24 N | |
COD Reagent | |
COD water determination |
Safety and Handling
- COD reagent acc. UNE 77-004-89
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- Carcinogenicity Category 1
- Substances and mixtures corrosive to metals Category 1
- Germ cell mutagenicity Category 1
- Reproductive toxicity Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360-May damage fertility or the unborn child.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : mercury(II) sulphate, potassium dichromate, sulphuric acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.