Learn More
Kern Sohn Compact Laboratory Balance PFB
Quick-display precision balance with user-friendly concept of operation - now with larger housing for more secure footing.
Brand:  Kern Sohn PFB 600-2
Encompass_Preferred
 View more versions of this product
Product Code. 17301723
  • 3602.67 SEK / Each
Explore more special offers
Description
Description
  • Easy to use: All primary functions have their own key on the keypad
  • Compact size, practical for small spaces
  • Capacity display: A bar graph display lights up to show how much of the weighing range is still available
  • Level indicator and levelling feet for precise levelling of the scale, fitted as standard, to give the most accurate weighing result
Specifications
Specifications
600 g
0.03 g
120 mm
Universal Mains Adapter
0.01 g
Compact Precision Balance
120 mm
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.