missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement factor H Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12748-1-AP-20UL
This item is not returnable.
View return policy
Description
Factor H is a regulatory factor of the alternative pathway of complement activation. Factor H is a glycoprotein consisting of a single polypeptide chain with a molecular mass of 150 kDa.Specifications
Complement factor H | |
Polyclonal | |
Unconjugated | |
CFH | |
ARMD4, ARMS1, CFH, CFHL3, Complement factor H, Factor H, FH, FHL1, H factor 1, HF, HF1, HF2, HUS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3075 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P08603 | |
CFH | |
Complement factor H Fusion Protein Ag3482 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction