missing translation for 'onlineSavingsMsg'
Learn More

Complement factor H Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen Complement factor H
Concentration 0.35 mg/mL
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16810634
View Documents
Proteintech
12748-1-AP-20UL
20 μL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16800634
View Documents
Proteintech
12748-1-AP-150UL
150 μL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Factor H is a regulatory factor of the alternative pathway of complement activation. Factor H is a glycoprotein consisting of a single polypeptide chain with a molecular mass of 150 kDa.
Specifications

Specifications

Complement factor H
Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Human
P08603
3075
Complement factor H Fusion Protein Ag3482
Primary
-20°C
CFH
0.35 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
ARMD4, ARMS1, CFH, CFHL3, Complement factor H, Factor H, FH, FHL1, H factor 1, HF, HF1, HF2, HUS
CFH
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.