missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CROP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14504-1-AP-150UL
This item is not returnable.
View return policy
Description
This is a rabbit polyclonal antibody raised against part of N-terminus of CROP of human origin and can recognize a ∽58 kDa band of CROP.
Binds cAMP regulatory element DNA sequence. May play a role in RNA splicing.Specifications
CROP | |
Polyclonal | |
Unconjugated | |
LUC7L3 | |
CRE associated protein 1, CREAP 1, CREAP1, CROP, Luc7 like protein 3, LUC7A, LUC7L3, O48, OA48 18 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51747, 67684 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95232, Q5SUF2 | |
LUC7L3 | |
CROP Fusion Protein Ag5961 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction