missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CROP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | CROP |
---|---|
Concentration | 0.33 mg/mL |
Applications | Western Blot, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16813174
|
Proteintech
14504-1-AP-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16803174
|
Proteintech
14504-1-AP-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This is a rabbit polyclonal antibody raised against part of N-terminus of CROP of human origin and can recognize a ∽58 kDa band of CROP.
Binds cAMP regulatory element DNA sequence. May play a role in RNA splicing.Specifications
CROP | |
Western Blot, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O95232, Q5SUF2 | |
51747, 67684 | |
CROP Fusion Protein Ag5961 | |
Primary | |
-20°C | |
LUC7L3 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CRE associated protein 1, CREAP 1, CREAP1, CROP, Luc7 like protein 3, LUC7A, LUC7L3, O48, OA48 18 | |
LUC7L3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title