Learn More
Cytosine-β-D-arabinofuranoside hydrochloride, MP BIOMEDICALS
Cytosine-β-D-Arabinofuranoside Hydrochloride
Brand:  MP Biomedicals 02100071.1
 View more versions of this product
Product Code. 11398359
  • 718.00 SEK / 100mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
Application Notes:
  • It is used as an antineoplastic and antiviral agent.
Usage Statement:
  • Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Key Applications:
  • Selective inhibitor of DNA synthesis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nucleoside
White
69-74-9
100 mg
C9H14ClN3O5
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytosine-β-D-arabinofuranoside hydrochloride, MP BIOMEDICALS > Cytosine-β-D-arabinofuranoside hydrochloride, 100 mg