Learn More
Tocris Bioscience™ DAPI
Fluorescent DNA stain
Brand: Tocris Bioscience™ 5748/50
This item is not returnable.
View return policy
Description
DAPI is a fluorescent DNA stain/ dye. DAPI stain binds to AT-rich regions of DNA and fluorescence increases by approximately 20-fold when bound to double-stranded DNA. DAPI stain can be used in flow cytometry, DNA visualization and quantification in histochemistry/biochemistry, to assess apoptosis, and to visualize mouse embryos or fetal organs. DAPI stain can be used in fixed cells and is cell-permeable at high concentrations providing effective cell staining. It is reported to be more photostable than Hoechst 33342 (Cat. No. 5117). Em/Ex λ = 350/470 nm respectively.Specifications
2-[4-(Aminoiminomethyl)phenyl]-1 H-Indole-6-carboximidamide hydrochloride | |
50 mg | |
C16H15N50.2 HCl | |
0.95 |
28718-90-3 | |
Fluorescent DNA Stains | |
350.25 |