missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DAPI
Fluorescent DNA stain
1327.18 SEK - 4821.69 SEK
Specifications
Chemical Name or Material | 2-[4-(Aminoiminomethyl)phenyl]-1 H-Indole-6-carboximidamide hydrochloride |
---|---|
CAS | 28718-90-3 |
Target | Fluorescent DNA Stains |
Molecular Formula | C16H15N50.2 HCl |
Formula Weight | 350.25 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18770108
|
Tocris Bioscience™
5748/10 |
10 mg |
1327.18 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18760298
|
Tocris Bioscience™
5748/50 |
50 mg |
4821.69 SEK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fluorescent DNA stain. Binds to AT-rich regions of DNA. Cell permeable at high concentrations.Specifications
2-[4-(Aminoiminomethyl)phenyl]-1 H-Indole-6-carboximidamide hydrochloride | |
Fluorescent DNA Stains | |
350.25 |
28718-90-3 | |
C16H15N50.2 HCl | |
0.95 |
Safety and Handling
Recommended Storage : Store at -20°C