Learn More
Datelliptium chloride, MedChemExpress
MedChemExpress Datelliptium chloride is a DNA-intercalating agent derived from ellipticine, with anti-tumor activities.
Brand: Medchem Express HY-U00337
Description
Datelliptium chloride is a DNA-intercalating agent derived from ellipticine, with anti-tumor activities.Specifications
Datelliptium chloride | |
Red | |
C23H28ClN3O | |
H2O : 41.67 mg/mL (104.71 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (12.56 mM; Need ultrasonic) | |
397.94 | |
98.33% | |
Solid |
105118-14-7 | |
Research | |
10 mg | |
CC1=C2C(NC3=C2C=C(O)C=C3)=C(C)C4=CC=[N+](CCN(CC)CC)C=C41.[Cl-] | |
397.94 | |
Research | |
Cancer-programmed cell death |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only