Learn More
Datelliptium chloride, MedChemExpress
MedChemExpress Datelliptium chloride is a DNA-intercalating agent derived from ellipticine, with anti-tumor activities.
Brand:  Medchem Express HY-U00337
Encompass_Preferred
 View more versions of this product
Product Code. 16410359
  • 3376.65 SEK / 10mg
Explore more special offers
Description
Description
Datelliptium chloride is a DNA-intercalating agent derived from ellipticine, with anti-tumor activities.
Specifications
Specifications
Datelliptium chloride
Red
C23H28ClN3O
H2O : 41.67 mg/mL (104.71 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (12.56 mM; Need ultrasonic)
397.94
98.33%
Solid
105118-14-7
Research
10 mg
CC1=C2C(NC3=C2C=C(O)C=C3)=C(C)C4=CC=[N+](CCN(CC)CC)C=C41.[Cl-]
397.94
Research
Cancer-programmed cell death
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Datelliptium chloride, MedChemExpress