Learn More
Borer Chemie™ deconex 20 NS-x
Liquid cleaner for manual and automated cleaning of laboratory glassware and precious items. deconex 20 NS-x delivers residue-free results in non-foaming processes and features a high emulsifying and dispersing power.
Brand: Borer Chemie™ 519100.00-K10W
This item is not returnable.
View return policy
Description
- Concentrate for manual and automated cleaning
- Suitable for immersion baths, ultrasonic baths and washers
- Without surfactants, silicates, chlorine, EDTA, or NTA
- High yield, typical concentration: 20-50 mL/L
- Broad cleaning spectrum
- High swelling and dispersing capacity leads to high performance
- Rinses off without leaving residues
- For consistent and flawless results
- Technical support by the manufacturer
Specifications
Liquid | |
Alkalis, complexing agents, dispersing agent, sequestering agents | |
Canister | |
1 x 10 kg | |
8.1 L | |
10 kg |
∼11.9 (1% Solution) | |
Concentrate | |
5°C to 25°C | |
2.1 gal. | |
22 lb. |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- deconex 20 NS-x
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P406-Store in corrosive resistant/ container with a resistant inner liner.
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide, tripotassiumorthophosphate