missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borer Chemie™ deconex 20 NS-x
Liquid cleaner for manual and automated cleaning of laboratory glassware and precious items. deconex 20 NS-x delivers residue-free results in non-foaming processes and features a high emulsifying and dispersing power.
1520.00 SEK - 9310.00 SEK
Specifications
Form | Liquid |
---|---|
pH | ∼11.9 (1% Solution) |
Composition | Alkalis, complexing agents, dispersing agent, sequestering agents |
Concentration | Concentrate |
Container Type | Canister |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17406512
|
Borer Chemie™
519100.00-KK5W |
1 x 5 kg |
1520.00 SEK
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16400795
|
Borer Chemie™
519100.00-K10W |
1 x 10 kg |
2970.00 SEK
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17704059
|
Borer Chemie™
519100.00-K25W |
1 x 30 kg |
9310.00 SEK
30kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Concentrate for manual and automated cleaning
- Suitable for immersion baths, ultrasonic baths and washers
- Without surfactants, silicates, chlorine, EDTA, or NTA
- High yield, typical concentration: 20-50 mL/L
- Broad cleaning spectrum
- High swelling and dispersing capacity leads to high performance
- Rinses off without leaving residues
- For consistent and flawless results
- Technical support by the manufacturer
Specifications
Liquid | |
Alkalis, complexing agents, dispersing agent, sequestering agents | |
Canister |
∼11.9 (1% Solution) | |
Concentrate | |
5°C to 25°C |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- deconex 20 NS-x
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P406-Store in corrosive resistant/ container with a resistant inner liner.
Supplemental information
- MIXTURE LIST-Contains : TRIPOTASSIUM ORTHOPHOSPHATE, POTASSIUM HYDROXIDE
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title