Learn More
Borer Chemie™ deconex 20 NS-x
Liquid cleaner for manual and automated cleaning of laboratory glassware and precious items. deconex 20 NS-x delivers residue-free results in non-foaming processes and features a high emulsifying and dispersing power.
Brand:  Borer Chemie™ 519100.00-KK5W
 View more versions of this product
Product Code. 17406512
  • 1520.00 SEK / 5kg
Explore more special offers
Description
Description
  • Concentrate for manual and automated cleaning
  • Suitable for immersion baths, ultrasonic baths and washers
  • Without surfactants, silicates, chlorine, EDTA, or NTA
  • High yield, typical concentration: 20-50 mL/L
  • Broad cleaning spectrum
  • High swelling and dispersing capacity leads to high performance
  • Rinses off without leaving residues
  • For consistent and flawless results
  • Technical support by the manufacturer
Specifications
Specifications
Liquid
Alkalis, complexing agents, dispersing agent, sequestering agents
Canister
1 x 5 kg
4 L
5 kg
∼11.9 (1% Solution)
Concentrate
5°C to 25°C
1.1 gal.
11 lb.
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • deconex 20 NS-x
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
Supplemental information
  • MIXTURE LIST-Contains : TRIPOTASSIUM ORTHOPHOSPHATE, POTASSIUM HYDROXIDE