Learn More
Dehydrocorydaline chloride, MedChemExpress
MedChemExpress Dehydrocorydaline chloride (13-Methylpalmatine chloride) is an alkaloid that regulates protein expression of Bax, Bcl-2; activates caspase-7, caspase-8, and inactivates PARP. Dehydrocorydaline chloride elevates p38 MAPK activation. Anti-inflammatory and anti-cancer activities. Dehydrocorydaline chloride shows strong anti-malarial effects (IC50 =38 nM), and low cytotoxicity (cell viability > 90%) using P. falciparum 3D7 strain.
Brand: Medchem Express HY-N0674A

Description
Dehydrocorydaline chloride (13-Methylpalmatine chloride) is an alkaloid that regulates protein expression of Bax, Bcl-2Specifications
Dehydrocorydaline chloride | |
10605-03-5 | |
5 mg | |
C22H24ClNO4 | |
13-Methylpalmatine chloride | |
CC1=C(C=CC(OC)=C2OC)C2=C[N+]3=C1C4=CC(OC)=C(OC)C=C4CC3.[Cl-] | |
401.88 | |
Research |
Solid | |
Yellow | |
Research | |
Cancer-Kinase/protease | |
DMSO : 25 mg/mL (62.21 mM; Need ultrasonic) | |
401.88 | |
98.0% |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only