Learn More
Dehydrocorydaline chloride, MedChemExpress
MedChemExpress Dehydrocorydaline chloride (13-Methylpalmatine chloride) is an alkaloid that regulates protein expression of Bax, Bcl-2; activates caspase-7, caspase-8, and inactivates PARP. Dehydrocorydaline chloride elevates p38 MAPK activation. Anti-inflammatory and anti-cancer activities. Dehydrocorydaline chloride shows strong anti-malarial effects (IC50 =38 nM), and low cytotoxicity (cell viability > 90%) using P. falciparum 3D7 strain.

2256.28 SEK - 3221.05 SEK
Specifications
Chemical Name or Material | Dehydrocorydaline chloride |
---|---|
Physical Form | Solid |
CAS | 10605-03-5 |
Color | Yellow |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16499058
![]() |
Medchem Express
HY-N0674A |
10 mg |
3221.05 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16451478
![]() |
Medchem Express
HY-N0674A |
5 mg |
2256.28 SEK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dehydrocorydaline chloride (13-Methylpalmatine chloride) is an alkaloid that regulates protein expression of Bax, Bcl-2Specifications
Dehydrocorydaline chloride | |
10605-03-5 | |
Research | |
Cancer-Kinase/protease | |
DMSO : 25 mg/mL (62.21 mM; Need ultrasonic) | |
401.88 | |
98.0% |
Solid | |
Yellow | |
C22H24ClNO4 | |
13-Methylpalmatine chloride | |
CC1=C(C=CC(OC)=C2OC)C2=C[N+]3=C1C4=CC(OC)=C(OC)C=C4CC3.[Cl-] | |
401.88 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.