Learn More
STEMCELL Technologies Delphinidin (Chloride)
Inhibits EGFR and VEGFR
Brand: STEMCELL Technologies 74234
Additional Details : Weight : 0.05000kg
Description
- Delphinidin is an anthocyanidin found in pigmented fruits and vegetables with anti-inflammatory, anti-angiogenic, and anti-oxidant properties (Watson & Schönlau)
- It has been reported to be a potent inhibitor of epidermal growth factor receptor (EGFR; Meiers et al.) and vascular endothelial growth factor receptor (VEGFR) (Lamy et al.) This product is supplied as the chloride salt of the molecule
- Induces G2-M phase arrest in human prostate cancer PC3 cells, leading to apoptosis (Hafeez et al.)
Specifications
528-53-0 | |
5 mg | |
≥97% |
Delphinidol; Delphinidine; Ephdine | |
C15H11ClO7 |