missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Delphinidin (Chloride)
Inhibits EGFR and VEGFR
POA - 1291.95 SEK
Specifications
CAS | 528-53-0 |
---|---|
Synonym | Delphinidol; Delphinidine; Ephdine |
Molecular Formula | C15H11ClO7 |
Purity | ≥97% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17120352
|
STEMCELL Technologies
74232 |
1 mg |
1291.95 SEK
1mg |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17130352
|
STEMCELL Technologies
74234 |
5 mg |
POA
5mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
Description
- Delphinidin is an anthocyanidin found in pigmented fruits and vegetables with anti-inflammatory, anti-angiogenic, and anti-oxidant properties (Watson & Schönlau)
- It has been reported to be a potent inhibitor of epidermal growth factor receptor (EGFR; Meiers et al.) and vascular endothelial growth factor receptor (VEGFR) (Lamy et al.) This product is supplied as the chloride salt of the molecule
- Induces G2-M phase arrest in human prostate cancer PC3 cells, leading to apoptosis (Hafeez et al.)
Specifications
528-53-0 | |
Delphinidol; Delphinidine; Ephdine | |
C15H11ClO7 | |
≥97% |