missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diallyl trisulfide, 98.8%, MP Biomedicals™
Chemical Identifiers
CAS | 2050-87-5 |
---|---|
Molecular Formula | C6H10S3 |
Molecular Weight (g/mol) | 178.326 |
InChI Key | UBAXRAHSPKWNCX-UHFFFAOYSA-N |
Synonym | diallyl trisulfide, allyl trisulfide, allitridin, trisulfide, di-2-propenyl, diallyltrisulfide, diallyl trisulphide, allitridum, dasuansu, diallyltrisulfane, di-2-propenyl trisulfide |
PubChem CID | 16315 |
ChEBI | CHEBI:78492 |
IUPAC Name | 3-(prop-2-enyltrisulfanyl)prop-1-ene |
SMILES | C=CCSSSCC=C |
Description
Chemical Identifiers
2050-87-5 | |
178.326 | |
diallyl trisulfide, allyl trisulfide, allitridin, trisulfide, di-2-propenyl, diallyltrisulfide, diallyl trisulphide, allitridum, dasuansu, diallyltrisulfane, di-2-propenyl trisulfide | |
CHEBI:78492 | |
C=CCSSSCC=C |
C6H10S3 | |
UBAXRAHSPKWNCX-UHFFFAOYSA-N | |
16315 | |
3-(prop-2-enyltrisulfanyl)prop-1-ene |
Specifications
2050-87-5 | |
0.988 | |
diallyl trisulfide, allyl trisulfide, allitridin, trisulfide, di-2-propenyl, diallyltrisulfide, diallyl trisulphide, allitridum, dasuansu, diallyltrisulfane, di-2-propenyl trisulfide | |
C=CCSSSCC=C | |
178.326 | |
CHEBI:78492 | |
Liquid |
Colorless | |
C6H10S3 | |
UBAXRAHSPKWNCX-UHFFFAOYSA-N | |
3-(prop-2-enyltrisulfanyl)prop-1-ene | |
16315 | |
178.3 | |
DATS |