missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Digi-Sense Replacement Calibration Salt
Digi-Sense Replacement Calibration Salt is designed especially for hygrometers.
Brand: Fisherbrand™ 35612-91
This item is not returnable.
View return policy
Description
- Keep your hygrometer running smoothly with these replacement items
Specifications
Sodium chloride | |
1 Ea. | |
Colorless | |
Hygrometer | |
NaCL | |
Soluble (358 g/L at 20°C) | |
75% RH salt | |
801°C | |
Hygrometer Calibration | |
Solid |
Bottle | |
Calibration Standard | |
Calibration Salt | |
7647-14-5 | |
Salt | |
58.44 g/mol | |
7 | |
1.413°C | |
Soluble |