missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Digi-Sense Replacement Calibration Salt
Digi-Sense Replacement Calibration Salt is designed especially for hygrometers.
640.00 SEK - 650.00 SEK
Specifications
Quantity | 20 g |
---|---|
Type | Calibration Salt |
For Use With (Equipment) | Traceable™ Hygrometers |
For Use With (Application) | Hygrometer Calibration |
Physical Form | Solid |
Product Code | Brand | dimPercentPurity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | dimPercentPurity | Price | Quantity & Availability | |||||
11765998
|
Fisherbrand™
35612-90 |
650.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11726008
|
Fisherbrand™
35612-91 |
640.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
- Keep your hygrometer running smoothly with these replacement items
Specifications
20 g | |
Traceable™ Hygrometers | |
Solid |
Calibration Salt | |
Hygrometer Calibration |