Learn More
OHAUS™ Dual Adjust Multi-Purpose Clamps
OHAUS™ Dual Adjust Multi-Purpose Clamps are designed to securely grip and position a variety of laboratory glassware.
Brand:  OHAUS™ 30392193
 View more versions of this product
Product Code. 15772239
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Extension arms with a 360° rotation allow placement of apparatus at various distances from lab-frame without compromising experiment integrity
  • Swivel and fixed position clamps hold apparatus near the lab-frame or stand, and have a built-in integral holder for attaching to lab-frames or apparatus
  • Available as single or dual-adjust in nickel-plated zinc or stainless steel
  • Non-slip vinyl sleeves, and fiberglass covers for temperatures above 100°C (included with all 2 and 3-prong clamps)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Multi-purpose Clamp
Non-slip vinyl sleeves and fiberglass covers
13 mm
9.020 in.
229 mm
4-Prong Dual Adjust
Nickel-plated
Clamp, Multi Purpose, CLM-HDCLP4DZS
14.00 in.
356.00 mm
24/40 mm Joint Size