Learn More
ABEBA™ Dynamic Flow Black Occupational Slip-On O1 ESD
Black dynamic flow occupational slip-on shoes with replaceable active MediFit insole, slip resistant TPU outsole
Brand: ABEBA™ 37376/44
This item is not returnable.
View return policy
Description
- Breathable, lightweight, slip-on occupational shoes
- Comply to EN ISO 20347: 2012, O1 SRC
- ESD dissipative according EN 61340 class climatic 1
- Slip resistant TPU outsole
- Good cushioning
- Energy absorption in heel area
- Black knitted fabric
- Double elastic band fitting system
- Removable active Medifit insole
Specifications
Black | |
44 | |
Unisex | |
Water Repellent |
Knitted Fabric | |
Comply to EN ISO 20347: 2012 O1 SRC, ESD class climatic 1 according EN 61340 | |
Slip-on | |
O1 SRC ESD |