Learn More
ABEBA™ Dynamic Flow Black Occupational Slip-On O1 ESD
Black dynamic flow occupational slip-on shoes with replaceable active MediFit insole, slip resistant TPU outsole
754.00 SEK
Specifications
Color | Black |
---|---|
Material | Knitted Fabric |
Certifications/Compliance (Europe) | Comply to EN ISO 20347: 2012 O1 SRC, ESD class climatic 1 according EN 61340 |
Gender | Unisex |
Ankle Style | Slip-on |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17121415
|
ABEBA™
37376/35 |
35 |
754.00 SEK
1 pair |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
17131415
|
ABEBA™
37376/36 |
36 |
754.00 SEK
1 pair |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
17121425
|
ABEBA™
37376/45 |
45 |
754.00 SEK
1 pair |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
17131425
|
ABEBA™
37376/46 |
46 |
754.00 SEK
1 pair |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
17141425
|
ABEBA™
37376/47 |
47 |
754.00 SEK
1 pair |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
17151425
|
ABEBA™
37376/48 |
48 |
754.00 SEK
1 pair |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
17141415
|
ABEBA™
37376/37 |
37 |
754.00 SEK
1 pair |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
17151415
|
ABEBA™
37376/38 |
38 |
754.00 SEK
1 pair |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
17161415
|
ABEBA™
37376/39 |
39 |
754.00 SEK
1 pair |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
17171415
|
ABEBA™
37376/40 |
40 |
754.00 SEK
1 pair |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
17181415
|
ABEBA™
37376/41 |
41 |
754.00 SEK
1 pair |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
17191415
|
ABEBA™
37376/42 |
42 |
754.00 SEK
1 pair |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
17101425
|
ABEBA™
37376/43 |
43 |
754.00 SEK
1 pair |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
17111425
|
ABEBA™
37376/44 |
44 |
754.00 SEK
1 pair |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
Description
- Breathable, lightweight, slip-on occupational shoes
- Comply to EN ISO 20347: 2012, O1 SRC
- ESD dissipative according EN 61340 class climatic 1
- Slip resistant TPU outsole
- Good cushioning
- Energy absorption in heel area
- Black knitted fabric
- Double elastic band fitting system
- Removable active Medifit insole
Specifications
Black | |
Comply to EN ISO 20347: 2012 O1 SRC, ESD class climatic 1 according EN 61340 | |
Slip-on | |
O1 SRC ESD |
Knitted Fabric | |
Unisex | |
Water Repellent |