Learn More
ABEBA™ Dynamic Flow Black Occupational Slip-On O1 ESD
Black dynamic flow occupational slip-on shoes with replaceable active MediFit insole, slip resistant TPU outsole
Brand:  ABEBA™ 37376/40
 View more versions of this product
Product Code. 17171415
  • 754.00 SEK / 1 pair
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Description
  • Breathable, lightweight, slip-on occupational shoes
  • Comply to EN ISO 20347: 2012, O1 SRC
  • ESD dissipative according EN 61340 class climatic 1
  • Slip resistant TPU outsole
  • Good cushioning
  • Energy absorption in heel area
  • Black knitted fabric
  • Double elastic band fitting system
  • Removable active Medifit insole
Specifications
Specifications
Black
40
Unisex
Water Repellent
Knitted Fabric
Comply to EN ISO 20347: 2012 O1 SRC, ESD class climatic 1 according EN 61340
Slip-on
O1 SRC ESD