Learn More
Labcon™ Eclipse™ 1200 μL Extended Length Pipette Tips for Sartorius™ Pipettes
These extended length pipette tips for Sartorius™ Pipettes come conveniently packed in resealable bags or racks.
Brand: Labcon™ 1048-960-006-9
This item is not returnable.
View return policy
Description
- Fits Sartorius™ 1200 μL Pipettes
- Low mounting and ejection forces less than 3 kg
- Polypropylene USP Class VI resin
- Made in USA
Specifications
1200 μL | |
Non-sterile | |
Autoclavable | |
ISO 9001:2015, FDA CFR21 GMP, USP Class VI | |
Micropipetting | |
96 Tips/Rack, 6 Racks/Pk., 8 Pk./Cs. | |
No | |
Polypropylene Resin |
Non-filtered | |
Clear | |
88.9 mm | |
ATP, RNase/Dnase, Endotoxin-free | |
Rack, 96 | |
Single-use | |
Eclipse |