Learn More
Eurofins DiscoverX EGFR (T790M, L858R) Protein, Active
Active, N-terminal GST-tagged, recombinant human EGFR, amino acids 696-end containing the mutations T790M and L858R, for use in Enzyme Assays.
Brand:  Eurofins DiscoverX 14-721M
Encompass_Preferred
 View more versions of this product
Product Code. 17310614
  • 46147.75 SEK / 250µg
Explore more special offers
This item is not returnable. View return policy
Description
Description
Receptors that are tyrosine protein kinases include those receptors for epidermal growth factor (EGF), platelet-derived growth factor (PDGF), insulin, and certain lymphokines. Receptor tyrosine kinases are cell surface transmembrane proteins responsible for intracellular signal transduction. They are expressed in many cell types and, after activation by growth factor binding, trigger a series of intracellular pathways leading to a wide variety of responses; e.g. differentiation, proliferation, migration and invasion, angiogenesis, survival. EGF receptor agonism may facilitate wound healing, enhance viral infections, topical inflammation and asthma as well as tumor cell proliferation and metastasis.
GenBank X00588
Specifications
Specifications
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
250 μg
Recombinant
Recombinant enzyme expressed in Sf21 insect cells
≥54%
85.8 kDa
Human
N-terminal GST
Truncated
EGFR (T790M,L858R), active
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Eurofins DiscoverX EGFR (T790M, L858R) Protein, Active