Learn More
Eurofins DiscoverX EGFR (T790M, L858R) Protein, Active
Active, N-terminal GST-tagged, recombinant human EGFR, amino acids 696-end containing the mutations T790M and L858R, for use in Enzyme Assays.

6046.30 SEK - 46147.75 SEK
Specifications
Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
---|---|
Molecular Weight (g/mol) | 85.8 kDa |
Species | Human |
Recombinant | Recombinant |
Protein Tag | N-terminal GST |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17300614
![]() |
Eurofins DiscoverX
14-721 |
10 μg |
6046.30 SEK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17310614
![]() |
Eurofins DiscoverX
14-721M |
250 μg |
46147.75 SEK
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Receptors that are tyrosine protein kinases include those receptors for epidermal growth factor (EGF), platelet-derived growth factor (PDGF), insulin, and certain lymphokines. Receptor tyrosine kinases are cell surface transmembrane proteins responsible for intracellular signal transduction. They are expressed in many cell types and, after activation by growth factor binding, trigger a series of intracellular pathways leading to a wide variety of responses; e.g. differentiation, proliferation, migration and invasion, angiogenesis, survival. EGF receptor agonism may facilitate wound healing, enhance viral infections, topical inflammation and asthma as well as tumor cell proliferation and metastasis.
GenBank X00588
Specifications
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. | |
Human | |
N-terminal GST | |
Truncated | |
EGFR (T790M,L858R), active |
85.8 kDa | |
Recombinant | |
Recombinant enzyme expressed in Sf21 insect cells | |
≥54% |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.