Learn More
Elma™ Elmasonic X-tra Flex 1 Ultrasonic Cleaner
Brand:  Elma™ 1033283
Additional Details : Weight : 45.00000kg
 View more versions of this product
Product Code. 11883142
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
Yes
3 x 400 VAC/N/PE
940 mm
3 x 400VAC/N/PE, 3650w
1200 mm
30°C to 80°C
90 kg
V2A Stainless Steel Casing, V4A Stainless Steel Tank
55 L
940 x 500 x 1200 mm
Industrial cleaning
Elmasonic X-tra Line 500 Flex 1, MF3O
3650 w
500 mm