missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-amino-4-(4-methoxyphenyl)-3-thiophenecarboxylate, 97%, Thermo Scientific™

365.42 SEK - 2700.95 SEK

Chemical Identifiers

CAS 15854-11-2
Molecular Formula C14H15NO3S
Molecular Weight (g/mol) 277.338
MDL Number MFCD01917484
InChI Key ZUOUKGJUHCKVDK-UHFFFAOYSA-N
Synonym ethyl 2-amino-4-4-methoxyphenyl thiophene-3-carboxylate, ethyl 2-amino-4-4-methoxyphenyl-3-thiophenecarboxylate, 2-amino-4-4-methoxyphenyl-thiophene-3-carboxylic acid ethyl ester, 2-amino-4-4-methoxyphenyl thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylicacid,2-amino-4-4-methoxyphenyl-,ethyl ester, 2-amino-4-4-methoxy-phenyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylicacid, 2-amino-4-4-methoxyphenyl-, ethyl esterShow MoreShow Less
PubChem CID 727641
IUPAC Name ethyl 2-amino-4-(4-methoxyphenyl)thiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC=C1C2=CC=C(C=C2)OC)N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10431283
SDS View Documents
Thermo Scientific Maybridge
MO07020DA
1 g Amber Glass Bottle
365.42 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
10380463
SDS View Documents
Thermo Scientific Maybridge
MO07020EA
10 g Amber Glass Bottle
2700.95 SEK
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ethyle2-amino-4-(4-methoxyphenyl)-3-thiophenecarboxylate, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.