Learn More
Emma Safety Footwear Everon S2 Shoe
Anatomically designed for a superior fit, the Everon is a shoe that's been designed for long days in and out of truck cabs, building sites, warehouses - wherever your work takes you.
Brand:  Emma Safety Footwear MM74854742
 View more versions of this product
Product Code. 17542556
  • 1365.00 SEK / 1 pair
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Description
  • S2-Safety shoe for wet working conditions
  • Metal Free
  • The energy returning Everon. Anatomically designed for a superior fit, the Everon is a shoe that's been designed for long days in and out of truck cabs, building sites, warehouses - wherever your work takes you.
  • The shoe's tongue has been ingeniously designed to prevent moisture and dirt entering.
  • Comfortable, supportive with energy returning soles, the Everon's a mid-boot version of the Evoke.
  • Industries:
    • Logistics & transport
Specifications
Specifications
Laces
Hydro-Tec Antibacterial
42
ISO 20345:2011
S2
Black, Gray
Nubuck
Reusable
Standard Width