Learn More
Exemestane, MedChemExpress
MedChemExpress Exemestane (FCE 24304) is a selective, irreversible and orally active steroidal aromatase inhibitor with IC50s of 30 nM and 40 nM for human placental and rat ovarian aromatase, respectively. Exemestane can be used for hormone-dependent breast cancer research.

1058.12 SEK - 2536.38 SEK
Specifications
Chemical Name or Material | Exemestane |
---|---|
Physical Form | Solid |
CAS | 107868-30-4 |
Color | White |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16471937
![]() |
Medchem Express
HY-13632 |
50 mg |
2038.43 SEK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16456256
![]() |
Medchem Express
HY-13632 |
100 mg |
2536.38 SEK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16415788
![]() |
Medchem Express
HY-13632 |
10 mg |
1058.12 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Exemestane (FCE 24304) is a selective, irreversible and orally active steroidal aromatase inhibitor with IC50s of 30 nM and 40 nM for human placental and rat ovarian aromatase, respectively. Exemestane can be used for hormone-dependent breast cancer research.Specifications
Exemestane | |
107868-30-4 | |
Research | |
Cancer-programmed cell death | |
DMSO : ≥ 54 mg/mL (182.19 mM) | |
296.4 | |
99.92% |
Solid | |
White | |
C20H24O2 | |
FCE 24304 EXE | |
C[C@]1([C@](CC2)([H])[C@]3([H])CC(C4=CC(C=C[C@]4(C)[C@@]3([H])CC1)=O)=C)C2=O | |
296.4 | |
Research |
Safety and Handling
H319∣H360
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.