Learn More
Lenz Laborglasintrumente Extraction Thimbles Porosity 0
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05405225
Additional Details : Weight : 0.50000kg
Description
- Extraction thimbles
- Glass insert with bores for solids
- Made of DURAN™ tubing
Specifications
30 mL | |
Borosilicate Glass (3.3) | |
0 |