Learn More
Fadrozole, MedChemExpress
MedChemExpress Fadrozole (CGS 16949A free base) is a potent, selective and nonsteroidal inhibitor of aromatase with an IC50 of 6.4 nM.
Brand: Medchem Express HY-14247A

Description
Fadrozole (CGS 16949A free base) is a potent, selective and nonsteroidal inhibitor of aromatase with an IC50 of 6.4 nM.Specifications
Fadrozole | |
Light Yellow | |
C14H13N3 | |
CGS 16949A free base (Rac)-FAD286 | |
N#CC1=CC=C(C2CCCC3=CN=CN23)C=C1 | |
223.27 | |
Research | |
Cancer-programmed cell death |
102676-47-1 | |
Research | |
50 mg | |
DMSO : ≥ 100 mg/mL (447.89 mM) | |
223.27 | |
99.34% | |
Solid |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only