Learn More
Fadrozole, MedChemExpress
MedChemExpress Fadrozole (CGS 16949A free base) is a potent, selective and nonsteroidal inhibitor of aromatase with an IC50 of 6.4 nM.

809.15 SEK - 7235.68 SEK
Specifications
Chemical Name or Material | Fadrozole |
---|---|
CAS | 102676-47-1 |
Color | Light Yellow |
Purity Grade Notes | Research |
Molecular Formula | C14H13N3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16496416
![]() |
Medchem Express
HY-14247A |
100 mg |
7235.68 SEK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16488617
![]() |
Medchem Express
HY-14247A |
5 mg |
809.15 SEK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16433416
![]() |
Medchem Express
HY-14247A |
25 mg |
2567.50 SEK
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16422147
![]() |
Medchem Express
HY-14247A |
50 mg |
4185.80 SEK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16406008
![]() |
Medchem Express
HY-14247A |
10 mg |
1291.53 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fadrozole (CGS 16949A free base) is a potent, selective and nonsteroidal inhibitor of aromatase with an IC50 of 6.4 nM.Specifications
Fadrozole | |
Light Yellow | |
C14H13N3 | |
DMSO : ≥ 100 mg/mL (447.89 mM) | |
223.27 | |
99.34% | |
Solid |
102676-47-1 | |
Research | |
CGS 16949A free base (Rac)-FAD286 | |
N#CC1=CC=C(C2CCCC3=CN=CN23)C=C1 | |
223.27 | |
Research | |
Cancer-programmed cell death |
Safety and Handling
H315∣H319∣H335
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.