Learn More
Ark Plas™ Female Bulkhead Lug Luer
Features conical tapers molded to ISO 594-1 and ISO 594-2 Standards
Brand:  Ark Plas™ LJX84-PP0
 View more versions of this product
Product Code. 15775835
  • 338.00 SEK / Pack of 25
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • All Ark-Plas Luer fittings feature conical tapers molded to ISO 594-1 and ISO 594-2 Standards for 6% luer tapers
  • This helps to ensure compatibility with all other luers molded to the same high standard as well as a broad variety of medical and laboratory instruments, including syringetips and needles or stopcocks and needles
  • All Luers are molded in virgin, lot-traceable material only
  • Specifications
    Specifications
    Polypropylene
    25
    Barb Size: 1/8