Learn More
Ark Plas™ Female Bulkhead Lug Luer
Features conical tapers molded to ISO 594-1 and ISO 594-2 Standards
337.00 SEK - 743.00 SEK
Specifications
Material Polypropylene
Packaging Quantity 25
View More Specs
Products 3
Product Code Brand Tubing Size Price Quantity & Availability  
Product Code Brand Tubing Size Price Quantity & Availability  
15775835
View Documents
Ark Plas™
LJX84-PP0
Barb Size: 1/8
338.00 SEK
Pack of 25
Add to basket
15765835
View Documents
Ark Plas™
LJX29-PP0
Barb Size: 1/16
337.00 SEK
Pack of 25
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
11821130
View Documents
Ark Plas™
LJX88-PP0
Barb Size: 3/32
743.00 SEK
Pack of 25
Estimated Shipment: 28-02-2025
to see stock.
Add to basket
Description
Description
  • All Ark-Plas Luer fittings feature conical tapers molded to ISO 594-1 and ISO 594-2 Standards for 6% luer tapers
  • This helps to ensure compatibility with all other luers molded to the same high standard as well as a broad variety of medical and laboratory instruments, including syringetips and needles or stopcocks and needles
  • All Luers are molded in virgin, lot-traceable material only
  • Specifications
    Specifications
    Polypropylene
    25