Learn More
Fendiline Hydrochloride
Inhibits KRas localization to the plasma membrane; also L-type calcium channel blocker
Brand:  Tocris 6407/50
Additional Details : Weight : 0.00970kg
 View more versions of this product
Product Code. 18775949
  • 1945.95 SEK / 50mg
Estimated Shipment: 09-09-2024
to see stock.
Description
Description
Fendiline Hydrochloride is a K-Ras inhibitor. Selectively inhibits K-Ras localization to the plasma membrane (IC50 = 9.64 μM). Blocks proliferation of cancer cell lines expressing oncogenic mutant K-Ras. Also inhibits L-type calcium channels (IC50 = 17 μM).
Specifications
Specifications
Ras GTPase Inhibitors
>98%
C23H25N.HCl
13636-18-5
50 mg
Observed MW: 351.91