missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fendiline Hydrochloride
Inhibits KRas localization to the plasma membrane; also L-type calcium channel blocker
582.06 SEK - 1945.95 SEK
Specifications
Chemical Name or Material | Ras GTPase Inhibitors |
---|---|
CAS | 13636-18-5 |
Assay Percent Range | >98% |
Molecular Formula | C23H25N.HCl |
Formula Weight | Observed MW: 351.91 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18798158
|
Tocris
6407/10 |
10 mg |
582.06 SEK
10mg |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18775949
|
Tocris
6407/50 |
50 mg |
1945.95 SEK
50mg |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
K-Ras inhibitor. Selectively inhibits K-Ras localization to the plasma membrane (IC50 = 9.64 μM). Blocks proliferation of cancer cell lines expressing oncogenic mutant K-Ras. Also inhibits L-type calcium channels (IC50 = 17 μM).Specifications
Ras GTPase Inhibitors | |
>98% | |
Observed MW: 351.91 |
13636-18-5 | |
C23H25N.HCl |