Learn More
Fendiline Hydrochloride
Inhibits KRas localization to the plasma membrane; also L-type calcium channel blocker
Brand: Tocris Bioscience 6407/50
This item is not returnable.
View return policy
Description
Fendiline Hydrochloride is a K-Ras inhibitor. Selectively inhibits K-Ras localization to the plasma membrane (IC50 = 9.64 μM). Blocks proliferation of cancer cell lines expressing oncogenic mutant K-Ras. Also inhibits L-type calcium channels (IC50 = 17 μM).Specifications
Ras GTPase Inhibitors | |
>98% | |
C23H25N.HCl |
13636-18-5 | |
50 mg | |
Observed MW: 351.91 |